Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAWR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006719436.3 | XP_006719499.1 | 5 | 3707 | ||
GenBank mRNA | AK027848.2 | - | 1 | 1208 |