Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB021923.1 | - | 1 | 100 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC029369.1 | - | 1 | 22 | Chr. 9 - 115924502 - 115925129 [-] on Build GRCh38 |