Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AJAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011541786.2 | XP_011540088.1 | 7 | 12110 | ||
GenBank mRNA | BC037321.2 | - | 1 | 2920 |