Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18orf63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174123.1 | NP_001167594.1 | 12 | 1742 | ||
GenBank mRNA | CR749350.1 | - | 12 | 1742 |