Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOBTB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017013251.1 | XP_016868740.1 | 1 | 1184 | ||
GenBank mRNA | AK092617.1 | - | 1 | 1184 |