Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TP53BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022533.1 | XP_016878022.1 | 14 | 3025 | ||
GenBank mRNA | DQ102772.1 | - | 1 | 117 |