Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542984.2 | XP_011541286.1 | 1 | 144 | ||
XM_017018305.1 | XP_016873794.1 | 1 | 144 | |||
XM_017018306.1 | XP_016873795.1 | 1 | 144 | |||
XM_017018308.1 | XP_016873797.1 | 1 | 144 | |||
XM_017018311.1 | XP_016873800.1 | 1 | 144 | |||
XM_017018312.1 | XP_016873801.1 | 1 | 144 | |||
GenBank mRNA | AF161533.1 | - | 1 | 576 |