Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001563.2 | NP_001001563.1 | 11 | 4152 | ||
GenBank mRNA | AK092396.1 | - | 1 | 137 | ||
AK097291.1 | - | 1 | 1396 | |||
BC014674.1 | - | 4 | 1484 |