Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATF7IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286514.1 | NP_001273443.1 | 15 | 7963 | ||
NM_018179.4 | NP_060649.3 | 15 | 7966 | |||
NM_181352.1 | NP_851997.1 | 15 | 8107 | |||
XM_005253424.3 | XP_005253481.1 | 15 | 8235 | |||
XM_006719108.3 | XP_006719171.1 | 15 | 8379 | |||
XM_006719109.3 | XP_006719172.1 | 15 | 8246 | |||
XM_011520754.2 | XP_011519056.1 | 15 | 7929 | |||
XM_011520755.2 | XP_011519057.1 | 15 | 8160 | |||
XM_017019638.1 | XP_016875127.1 | 15 | 7825 | |||
XM_017019639.1 | XP_016875128.1 | 15 | 8376 | |||
GenBank mRNA | AK125103.1 | - | 1 | 1559 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM719228.1 | - | 1 | 30 | Chr. 12 - 14501994 - 14502674 [+] on Build GRCh38 |