Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC018460.1 | - | 2 | 244 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG576414.1 | - | 2 | 282 | Chr. 12 - 29149312 - 29151863 [+] on Build GRCh38 |