Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHODH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005255827.3 | XP_005255884.1 | 9 | 4754 | ||
XM_005255829.3 | XP_005255886.1 | 8 | 4391 | |||
XM_017022990.1 | XP_016878479.1 | 9 | 4529 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK098239.1 | - | 3 | 2060 | Chr. 16 - 72025041 - 72027663 [+] on Build GRCh38 |