Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316973.1 | NP_001303902.1 | 8 | 2194 | ||
NM_001316974.1 | NP_001303903.1 | 8 | 2137 | |||
NM_001316976.1 | NP_001303905.1 | 6 | 2016 | |||
NM_032368.4 | NP_115744.2 | 8 | 2294 | |||
XM_005263509.2 | XP_005263566.1 | 6 | 2011 | |||
XM_017002546.1 | XP_016858035.1 | 8 | 2040 | |||
XM_017002547.1 | XP_016858036.1 | 8 | 2060 | |||
GenBank mRNA | AK025913.1 | - | 1 | 1391 |