Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRIP12 SIRNA Assays ›
Gene Symbol:
TRIP12
Gene Name:
thyroid hormone receptor interactor 12
Gene Aliases:
TRIP-12, ULF
Chromosome Location:
Chr.2: 229763837 - 229923234 on Build GRCh38
UniGene ID:
Hs.572642
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
n348627
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRIP12
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001284214.1
NP_001271143.1
42
8576
NM_001284215.1
NP_001271144.1
40
8401
NM_001284216.1
NP_001271145.1
39
7622
NM_004238.2
NP_004229.1
41
8432
XM_005246954.4
XP_005247011.1
42
8684
XM_005246955.4
XP_005247012.1
42
8683
XM_005246956.4
XP_005247013.1
42
8660
XM_005246957.4
XP_005247014.1
42
8635
XM_005246958.4
XP_005247015.1
42
8603
XM_005246960.4
XP_005247017.1
43
8597
XM_005246961.3
XP_005247018.1
41
8534
XM_005246962.4
XP_005247019.1
41
8531
XM_006712852.3
XP_006712915.1
41
8487
XM_011512180.2
XP_011510482.1
42
8637
XM_011512181.2
XP_011510483.1
42
8635
XM_011512183.2
XP_011510485.1
41
8491
XM_017005272.1
XP_016860761.1
43
8743
XM_017005273.1
XP_016860762.1
42
8610
XM_017005274.1
XP_016860763.1
42
8558
XM_017005275.1
XP_016860764.1
42
8613
XM_017005276.1
XP_016860765.1
42
8618
XM_017005277.1
XP_016860766.1
43
8758
XM_017005278.1
XP_016860767.1
42
8573
XM_017005279.1
XP_016860768.1
42
8630
XM_017005280.1
XP_016860769.1
42
8619
XM_017005281.1
XP_016860770.1
42
8553
XM_017005282.1
XP_016860771.1
43
8596
XM_017005283.1
XP_016860772.1
42
8585
XM_017005284.1
XP_016860773.1
43
8579
XM_017005285.1
XP_016860774.1
41
8521
XM_017005286.1
XP_016860775.1
41
8553
XM_017005287.1
XP_016860776.1
41
8519
XM_017005288.1
XP_016860777.1
41
8481
XM_017005289.1
XP_016860778.1
43
8516
XM_017005290.1
XP_016860779.1
43
8513
XM_017005291.1
XP_016860780.1
42
8447
XM_017005292.1
XP_016860781.1
41
8435
XM_017005293.1
XP_016860782.1
41
8450
GenBank mRNA
AK128481.1
-
1
1462
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
DNA repair
cellular response to DNA damage stimulus
embryo development
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of histone H2A K63-linked ubiquitination
negative regulation of double-strand break repair
ubiquitin-protein transferase activity
protein binding
ligase activity
thyroid hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0