Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC729224 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135244.1 | 2 | 466 | |||
GenBank mRNA | BC040292.1 | - | 1 | 1454 | ||
DB055974.1 | - | 2 | 420 |