Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC017429.1 | - | 1 | 265 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG566108.1 | - | 1 | 262 | Chr. 12 - 50108101 - 50108912 [+] on Build GRCh38 |