Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271572.1 | NP_001258501.1 | 3 | 1638 | ||
NM_001271573.1 | NP_001258502.1 | 2 | 1615 | |||
NM_003847.2 | NP_003838.1 | 3 | 1731 | |||
GenBank mRNA | AL360141.1 | - | 1 | 845 |