Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270975.1 | NP_001257904.1 | 10 | 2894 | ||
NM_001270976.1 | NP_001257905.1 | 11 | 3009 | |||
NM_001270977.1 | NP_001257906.1 | 9 | 2801 | |||
NM_001270978.1 | NP_001257907.1 | 7 | 2522 | |||
NM_001270979.1 | NP_001257908.1 | 6 | 2438 | |||
NM_014761.3 | NP_055576.2 | 10 | 2890 | |||
GenBank mRNA | BC038350.1 | - | 1 | 408 |