Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGK494 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174103.1 | NP_001167574.1 | 12 | 2403 | ||
GenBank mRNA | AK056265.1 | - | 1 | 212 | ||
BC001565.1 | - | 1 | 1602 |