Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK226139.1 | - | 1 | 4765 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK024390.1 | - | 1 | 2060 | Chr. 17 - 45975490 - 45980506 [+] on Build GRCh38 |