Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTOV1-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110730.1 | 5 | 792 | |||
GenBank mRNA | AL050131.1 | - | 1 | 1610 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM992525.1 | - | 4 | 616 | Chr. 19 - 49856732 - 49860742 [-] on Build GRCh38 |