Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOCS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014598.3 | NP_055413.1 | 10 | 4429 | ||
XM_017024551.1 | XP_016880040.1 | 9 | 4575 | |||
XM_017024552.1 | XP_016880041.1 | 8 | 4470 | |||
GenBank mRNA | AK094338.1 | - | 1 | 1884 |