Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGI2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038343.2 | 4 | 2969 | |||
GenBank mRNA | AK021837.1 | - | 1 | 433 | ||
AL833456.1 | - | 2 | 2694 | |||
BX641113.1 | - | 1 | 1861 | |||
CR749529.1 | - | 2 | 3022 |