Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCM3AP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC030022.1 | - | 2 | 1652 |
Gene Symbol: MCM3AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA659734.1 | - | 1 | 33 |