Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128847.1 | - | 1 | 177 |
Gene Symbol: LOC100996274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011526289.2 | XP_011524591.1 | 1 | 177 |