Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC036416.1 | - | 1 | 633 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG707422.1 | - | 1 | 633 | Chr. 2 - 36851696 - 36852512 [-] on Build GRCh38 |