Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047649.1 | 12 | 1695 | |||
GenBank mRNA | BX641065.1 | - | 6 | 2566 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK057666.1 | - | 1 | 1337 | Chr. 5 - 55633021 - 55635083 [-] on Build GRCh38 |