Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK054677.1 | - | 5 | 1411 | ||
BC052958.1 | - | 3 | 2341 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ424231.1 | - | 1 | 340 | Chr. 7 - 56040373 - 56041201 [-] on Build GRCh38 |