Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017007985.1 | XP_016863474.1 | 5 | 8208 | ||
GenBank mRNA | AK124778.1 | - | 1 | 1330 |