Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MITF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017006444.1 | XP_016861933.1 | 1 | 7176 | ||
GenBank mRNA | AY632574.1 | - | 1 | 204 |