Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDFY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020386.1 | XP_016875875.1 | 2 | 4505 | ||
GenBank mRNA | AK054833.1 | - | 1 | 528 |