Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM185B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024121.2 | NP_077026.2 | 1 | 5903 | ||
GenBank mRNA | AK021716.1 | - | 2 | 1598 | ||
AK090980.1 | - | 1 | 2009 |