Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM63B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040450.2 | NP_001035540.1 | 9 | 6765 | ||
NM_001040453.2 | NP_001035543.1 | 9 | 6592 | |||
XM_011521687.1 | XP_011519989.1 | 10 | 6817 | |||
XM_011521688.1 | XP_011519990.1 | 10 | 6814 | |||
XM_011521689.1 | XP_011519991.1 | 10 | 6715 | |||
XM_011521690.1 | XP_011519992.1 | 9 | 6661 | |||
XM_017022332.1 | XP_016877821.1 | 8 | 6571 | |||
XM_017022333.1 | XP_016877822.1 | 7 | 6517 | |||
XM_017022334.1 | XP_016877823.1 | 7 | 6514 | |||
GenBank mRNA | AK021710.1 | - | 1 | 835 |