Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011523379.1 | XP_011521681.1 | 1 | 827 | ||
XM_011523381.1 | XP_011521683.1 | 1 | 827 | |||
GenBank mRNA | AK130680.1 | - | 1 | 740 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC035173.1 | - | 1 | 1539 | Chr. 16 - 89816769 - 89818757 [+] on Build GRCh38 |