Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033119.4 | NP_149110.1 | 10 | 7227 | ||
XM_011523399.2 | XP_011521701.1 | 13 | 7538 | |||
XM_011523400.2 | XP_011521702.1 | 8 | 6829 | |||
XM_017023804.1 | XP_016879293.1 | 12 | 8757 | |||
GenBank mRNA | AK021701.1 | - | 1 | 1504 | ||
AK023784.1 | - | 1 | 1779 |