Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFNA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI459151.1 | - | 1 | 56 | ||
AI625552.1 | - | 1 | 32 | |||
BC042610.1 | - | 1 | 2213 |