Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGCG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003358.2 | NP_003349.1 | 9 | 3515 | ||
XM_005252186.4 | XP_005252243.1 | 10 | 3538 | |||
GenBank mRNA | AJ420423.1 | - | 1 | 1237 |