Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005206.4 | NP_005197.3 | 3 | 2929 | ||
NM_016823.3 | NP_058431.2 | 3 | 3099 | |||
GenBank mRNA | AK095449.1 | - | 1 | 1409 |