Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933432G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040716.1 | 5 | 752 | |||
GenBank mRNA | AK017021.1 | - | 5 | 756 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY716705.1 | - | 5 | 752 | Chr. 9 - 118429839 - 118437039 [+] on Build GRCm38 |