Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933432G23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040716.1 | 5 | 826 | |||
GenBank mRNA | AK017021.1 | - | 5 | 830 |