Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Six3os1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038082.1 | 4 | 2480 | |||
NR_038083.1 | 4 | 1564 | ||||
GenBank mRNA | AK053722.1 | - | 4 | 1566 | ||
BC065087.1 | - | 3 | 3364 |