Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E230015B07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042791.1 | - | 3, 4 | 357 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB639992.1 | - | 3, 4 | 357 | Chr. 6 - 87672382 - 87679135 [+] on Build GRCm38 |