Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm20658 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ308964.1 | - | 2 | 150 | ||
AK080151.1 | - | 4 | 1429 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB221816.2 | - | 2 | 143 | Chr. 12 - 116102792 - 116110680 [-] on Build GRCm38 |