Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1700102F20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007111.2 | - | 1 | 647 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707187.1 | - | 1 | 649 | Chr. 6 - 140309071 - 140309740 [+] on Build GRCm38 |