Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK076423.1 | - | 19 | 3708 | ||
AK077756.1 | - | 1 | 374 | |||
BC127081.1 | - | 13 | 2842 |