Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm2861 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK080443.1 | - | 473 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB639830.1 | - | 2 | 467 | Chr. 5 - 97195591 - 97207922 [+] on Build GRCm38 |