Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slx1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033446.1 | 5 | 2788 | |||
GenBank mRNA | AK003973.1 | - | 1 | 171 | ||
AK153779.1 | - | 1 | 770 | |||
AK163709.1 | - | 1 | 598 |