Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5730488B01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073462.1 | 1 | 2080 | |||
GenBank mRNA | AK017710.1 | - | 1 | 2081 |