Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E230032D23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK037811.1 | - | 1 | 363 | ||
AK054221.1 | - | 1 | 313 | |||
AK154738.1 | - | 1 | 108 | |||
BB160234.2 | - | 1 | 309 |
Gene Symbol: AK155734 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK155734.1 | - | 1 | 106 |