Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Azin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007010.1 | - | 896 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX516018.1 | - | 1 | 247 | Chr. 15 - 38519438 - 38519940 [+] on Build GRCm38 |