Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3gap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005623.1 | - | 1 | 270 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY705821.1 | - | 1 | 270 | Chr. 1 - 127938379 - 127938882 [-] on Build GRCm38 |